Skip to Content
Merck
All Photos(1)

Key Documents

04-639

Sigma-Aldrich

Anti-PP2A Antibody, B subunit, B56 delta, clone H5D12

clone H5D12, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€382.00

€382.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€382.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€382.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

H5D12, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP2R5D(5528)

General description

PP2A is a heterotrimeric phosphatase, ccomposed of A, B and C subunits. The structural A and catalytic C subunits form the constitutive core of the enzyme, which associates with one of a large number of regulatory B subunits. Three families of B subunits have been identified thus far. The prototypical B (or PR55) family of subunits is encoded by three genes, Balpha, Bbeta and Bgamma . The B′ (or B56) family consists of at least seven splice variantsderived from five genes. RNA′s for the two members of the B′′ family, PR72 and PR130, arise by alternative splicing or use of alternate transcription start sites from the same gene. Primary structures as well as functions of B and B′ subunits are conserved from yeast to mammals.

Specificity

Human PP2A, B subunit, B56 delta. This monoclonal antibody is the delta isoform for the B56 family of protein phosphatase 2A.

Immunogen

GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta

Application

Immunoblotting (1:500 to 1:2000)

Optimal working dilutions must be determined by the end user
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases
This Anti-PP2A Antibody, B subunit, B56 delta, clone H5D12 is validated for use in WB for the detection of PP2A.

Physical form

Format: Purified
Purified mouse monoclonal IgG1 antibody in PBS with 0.05% NaN3.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John D Lapek et al.
Molecular & cellular proteomics : MCP, 16(8), 1447-1461 (2017-06-14)
The mechanisms by which human immunodeficiency virus (HIV) circumvents and coopts cellular machinery to replicate and persist in cells are not fully understood. HIV accessory proteins play key roles in the HIV life cycle by altering host pathways that are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service