Skip to Content
Merck
All Photos(2)

Key Documents

04-282

Sigma-Aldrich

Anti-phospho-EGFR (Thr654) Antibody, clone 3F2

clone 3F2, Upstate®, from mouse

Synonym(s):

ERBB, ERBB1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€389.00

€389.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€389.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€389.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F2, monoclonal

species reactivity

mouse, human, canine, rat

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr654)

Gene Information

dog ... Egfr(404306)
human ... EGFR(1956)
mouse ... Egfr(13649)
rat ... Egf(25313)

Specificity

Recognizes phosphorylated EGFR on Threonine 654

Immunogen

KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Thr 654 in human EGFR

Application

Detect phospho-EGFR (Thr654) using this Anti-phospho-EGFR (Thr654) Antibody, clone 3F2 validated for use in ELISA, IP & WB.

Quality

Routinely evaluated by immunoblot.

Target description

180 kDa

Physical form

100 µg (0.1 mg/ml) of mouse monoclonal IgG1 in PBS containing 0.09% sodium azide, PEG, sucrose and 50% glycerol
Format: Purified

Analysis Note

Control
Includes PMA and pervanadate treated Hep2G cell lysate as a positive control

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation of Thr654 but not Thr669 within the juxtamembrane domain of the EGF receptor inhibits calmodulin binding
Aifa, Sami, et al
Biochemical and biophysical research communications, 347, 381-387 (2006)
Protein kinase C phosphorylation of the EGF receptor at a threonine residue close to the cytoplasmic face of the plasma membrane
Hunter, T, et al
Nature, 311, 480-483 null
Elena Odintsova et al.
The Journal of biological chemistry, 288(36), 26323-26334 (2013-07-31)
Ligand-induced ubiquitylation of EGF receptor (EGFR) is an important regulatory mechanism that controls endocytic trafficking of the receptor and its signaling potential. Here we report that tetraspanin CD82/KAI1 specifically suppresses ubiquitylation of EGFR after stimulation with heparin-binding EGF or amphiregulin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service