Skip to Content
Merck
All Photos(1)

Key Documents

T-907

Supelco

Temazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Temazepam Standard Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN2O2
Molecular Weight:
300.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CN1C(=O)C(O)N=C(c2ccccc2)c3cc(Cl)ccc13

InChI

1S/C16H13ClN2O2/c1-19-13-8-7-11(17)9-12(13)14(18-15(20)16(19)21)10-5-3-2-4-6-10/h2-9,15,20H,1H3

InChI key

SEQDDYPDSLOBDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sold under the trade names Restoril® and Normison, temazepam is a benzodiazepine approved for the short term treatment of insomnia. The drug has anti-anxiety, anticonvulsant and skeletal muscle relaxant properties. This certified Snap-N-Spike® solution is suitable for a variety of GC/MS and LC/MS testing applications from forensic analysis and pain prescription monitoring to clinical toxicology and urine drug testing.

Application


  • Evaluation of an activated carbon disposal system: This study evaluates an activated carbon disposal system for safely disposing of model prescription sedative medications, including Temazepam, focusing on the environmental aspects and the prevention of medication abuse (Dasht Bozorg et al., 2020).

  • Rapid determination of benzodiazepines in pharmaceuticals: Research on the rapid determination of benzodiazepines, including Temazepam, using surface-enhanced Raman scattering demonstrates potential applications in the identification of counterfeit pharmaceuticals (Kimani et al., 2021).

  • Biologically active benzodiazepine metabolites: A study on the formation of biologically active benzodiazepine metabolites in hydroponic conditions used Arabidopsis thaliana cell cultures to explore environmental and pharmacological impacts, relevant to Temazepam research (Dudley et al., 2019).

  • Racemization of chiral 1,4-benzodiazepines: Investigation into the racemization of oxazepam and other 1,4-benzodiazepines in aqueous solution provides insights into the stability and behavior of these compounds, which includes Temazepam, under laboratory conditions (Hok et al., 2019).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Restoril is a registered trademark of Mallinckrodt Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service