Skip to Content
Merck
All Photos(1)

Key Documents

NMID773C

Supelco

Testosterone Enanthate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59
UNSPSC Code:
41116107

grade

certified reference material

Agency

NMI Australia certified reference material

form

solid

manufacturer/tradename

NMI Australia

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

2-8°C

InChI

1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1

InChI key

VOCBWIIFXDYGNZ-IXKNJLPQSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service