Skip to Content
Merck
All Photos(1)

Key Documents

D-088

Supelco

N-Desmethylcyclobenzaprine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€161.00

€161.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€161.00

About This Item

Empirical Formula (Hill Notation):
C19H19N · HCl
CAS Number:
Molecular Weight:
297.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€161.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CNCCC=C1C2=C(C=CC=C2)C=CC3=C1C=CC=C3.Cl

InChI

1S/C19H19N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-13,20H,6,14H2,1H3;1H

InChI key

UZMPCPFDZYTEJG-UHFFFAOYSA-N

General description

A certified Snap-N-Spike® Solution suitable for cyclobenzaprine testing methods by GC/MS or LC/MS for applications in pain prescription monitoring, forensic analysis, urine drug testing, or clinical toxicology. Also known as norcyclobenzaprine, N-desmethylcyclobenzaprine is the major urinary metabolite of the skeletal muscle relaxant cyclobenzaprine. Cyclobenzaprine, sold under trade names Flexeril®, Fexmid®, and Amrix®, is used to relieve skeletal muscle spasms and associated pain from acute musculoskeletal conditions.

Legal Information

Amrix is a registered trademark of Ivax International GMBH LLC
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Fexmid is a registered trademark of Shionogi Inc.
Flexeril is a registered trademark of ALZA Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service