Skip to Content
Merck
All Photos(1)

Documents

M-171

Supelco

3-Methoxytyramine hydrochloride

1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Synonym(s):

3-MT, 3-Methoxy-4-hydroxyphenethylamine, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3-4-(OH)CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
3631283
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H

InChI key

AWRIOTVUTPLWLF-UHFFFAOYSA-N

General description

3-Methoxytyramine is a metabolite of dopamine, a catecholamine which acts as a neurotransmitter for dopamine receptors and a neurohormone for regulation of numerous brain functions. 3-Methoxytyramine levels can serve as a biomarker for the diagnosis of neuroendocrine tumors such as pheochromocytoma or paraganglioma. This certified solution standard is suitable for LC/MS applications in diagnostic testing, endocrinology, and clinical chemistry.

Application


  • Dopamine metabolite analysis: 3-Methoxytyramine HCl is integral to advanced methods for sensitive and simultaneous quantification of neurotransmitters and their metabolites, enhancing our understanding of neurochemical pathways in various neurological disorders (Becker et al., 2023).

  • Neurotransmitter research: Utilized in cutting-edge metabolic profiling techniques such as matrix-assisted laser desorption/ionization mass spectrometry imaging, 3-Methoxytyramine HCl helps differentiate neurological conditions(Vallianatou et al., 2023).

  • Catecholamine biochemistry: In research for diagnosing pheochromocytomas and paragangliomas, 3-Methoxytyramine HCl is used to develop dual-template magnetic molecularly imprinted polymers for robust detection of urine metanephrines, demonstrating its critical role in medical diagnostics (Zeng et al., 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service