Skip to Content
Merck
All Photos(1)

Key Documents

E-077

Supelco

(±)-Epinephrine-D6 solution

100 μg/mL in methanol with 5% 1 M HCl, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-Epinephrine-D6

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€188.00

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€188.00

About This Item

Empirical Formula (Hill Notation):
C9H7D6NO3
CAS Number:
Molecular Weight:
189.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 5% 1 M HCl

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

N(C(C(O)(c1c(c(c(c(c1[2H])[2H])O)O)[2H])[2H])([2H])[2H])C

InChI

1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/i2D,3D,4D,5D2,9D

InChI key

UCTWMZQNUQWSLP-JQAOAMCHSA-N

General description

Epinephrine, also known as adrenaline, is a marker for catecholamine-secreting cancers such as pheochromocytoma, paraganglioma, and neuroblastoma. This internal standard is suitable for LC-MS/MS monitoring of epinephrine levels in urine or plasma for diagnostic testing or endocrinology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service