Skip to Content
Merck
All Photos(1)

Key Documents

E-067

Supelco

Ethosuximide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO2
CAS Number:
Molecular Weight:
141.17
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

CCC1(C)CC(=O)NC1=O

InChI

1S/C7H11NO2/c1-3-7(2)4-5(9)8-6(7)10/h3-4H2,1-2H3,(H,8,9,10)

InChI key

HAPOVYFOVVWLRS-UHFFFAOYSA-N

Gene Information

General description

Ethosuximide is an anticonvulsant specifically used to treat absence seizures. This Snap-N-Spike® Reference Solution is suitable for LC/MS or GC/MS applications in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dema M Ajwee et al.
Chemical biology & drug design, 79(1), 137-142 (2011-02-22)
The fact that ethosuximide (ETO), phenobarbital (PHO), and barbituric acid (BARB) share structural and pharmacophoric homologies with phenytoin and allantoin, both known to have significant wound-healing properties, prompted us to evaluate them as wound-healing agents. Accordingly, ETO-, PHO-, and BARB-containing
Gilles van Luijtelaar et al.
International journal of psychophysiology : official journal of the International Organization of Psychophysiology, 85(1), 49-54 (2011-09-29)
Recently it was established that early long lasting treatment with the anti-absence drug ethosuximide (ETX) delays the occurrence of absences and reduces depressive-like symptoms in a genetic model for absence epilepsy, rats of the WAG/Rij strain. Here it is investigated
Emilio Russo et al.
Epilepsia, 52(7), 1341-1350 (2011-06-04)
Depression is most commonly associated with epilepsy. Recent reports have suggested a putative relationship between seizure development and onset of depressive behavior, whereas others proposed that two clinical entities might represent different neuropathologic aspects of the same neurologic disorder. The
Paolo Bonanni et al.
Developmental medicine and child neurology, 54(10), 961-964 (2012-03-15)
At 7 years of age, a female with mucopolysaccharidosis type II (MPS II) showed a sudden deterioration in neurological function, a sleep disorder, and progressive behavioural impairment. Electroencephalography was performed 1 year and 8 months after the onset of the
Gabi Dezsi et al.
Epilepsia, 54(4), 635-643 (2013-03-08)
Ethosuximide (ESX) is a drug of choice for the symptomatic treatment of absence seizures. Chronic treatment with ESX has been reported to have disease-modifying antiepileptogenic activity in the WAG/Rij rat model of genetic generalized epilepsy (GGE) with absence seizures. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service