Skip to Content
Merck
All Photos(1)

Key Documents

B-041

Supelco

Barbiturate Mix-5 solution

250 μg/mL each component in methanol, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product B-041 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Amobarbital
Butalbital
Pentobarbital
Phenobarbital
Secobarbital

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC/IV (Portugal)

concentration

250 μg/mL each component in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A certified multi-component solution mix of routinely tested barbiturates amobarbital, butalbital, pentobarbital, phenobarbital and secobarbital. Barbiturates are drugs that act as CNS depressants, and therefore produce a wide range of effects such as sedation and anesthesia. This certified solution mix is suitable for GC/MS and LC/MS applications in forensic analysis, clinical toxicology, and urine drug testing.

Application

  • Barbiturate reference standards: Although specific PubMed articles are not available, Barbiturate Mix-5 solution is a crucial analytical tool in the pharmaceutical sector, used as a reference standard for the calibration and validation of analytical instrumentation, particularly in chromatography, to ensure the precise quantification of barbiturates in drug formulations and forensic samples.
  • CNS depressant research: This mix is vital for biochemical assays that explore the pharmacodynamics and pharmacokinetics of central nervous system depressants, aiding in the understanding of their therapeutic and side effect profiles.
  • Pharmaceutical compound calibration: Barbiturate Mix-5 solution serves as an essential calibration standard in pharmaceutical research, helping to maintain stringent quality control measures and compliance with regulatory standards for drug safety and efficacy.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service