Skip to Content
Merck
All Photos(1)

Key Documents

999971P

Avanti

1-C16 Ether MG

Avanti Research - A Croda Brand 999971P, powder

Synonym(s):

1-O-hexadecyl-sn-glycerol (HG)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€311.00

€311.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€311.00

About This Item

Empirical Formula (Hill Notation):
C19H40O3
CAS Number:
Molecular Weight:
316.52
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€311.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 250 mg (999971P-250mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999971P

lipid type

phospholipids
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](CO)(O)COCCCCCCCCCCCCCCCC

InChI

1S/C19H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-22-18-19(21)17-20/h19-21H,2-18H2,1H3

InChI key

OOWQBDFWEXAXPB-UHFFFAOYSA-N

General description

1-C16 Ether MG or 1-O-hexadecyl-sn-glycerol (HG) is a neutral lipid containing a saturated 16-carbon alkyl chain ether and an uncharged headgroup.[1]

Application

1-C16 Ether MG or 1-O-hexadecyl-sn-glycerol (HG) might be used to study its effect on target gene expression in HepG2 cells.[2] It might also be used to promote plasmalogen biosynthesis in lymphoblasts derived from Barth syndrome (BTHS) patients.[1]

Biochem/physiol Actions

1-C16 Ether MG or 1-O-hexadecyl-sn-glycerol (HG) acts as a precursor for plasma plasmalogen (PI) biosynthesis.[2] It is essential for maintaining the plasmalogen levels in cell lines.[3]

Packaging

20 mL Clear Glass Screw Cap Vial (999971P-250mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh Phuyal et al.
The Journal of biological chemistry, 290(7), 4225-4237 (2014-12-19)
Exosomes are vesicles released by cells after fusion of multivesicular bodies with the plasma membrane. In this study, we have investigated whether ether lipids affect the release of exosomes in PC-3 cells. To increase the cellular levels of ether lipids
Extrinsic effectors regulating genes for plasmalogen biosynthetic enzymes in HepG2 cells
Maeba R, et al.
Biomedical Research and Clinical Practice, 2 (2017)
Sara L Morales-Lázaro et al.
The Journal of biological chemistry, 289(35), 24079-24090 (2014-07-19)
The transient receptor potential vanilloid 1 (TRPV1) ion channel is a polymodal protein that responds to various stimuli, including capsaicin (the pungent compound found in chili peppers), extracellular acid, and basic intracellular pH, temperatures close to 42 °C, and several
José Carlos Bozelli et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(6), 158677-158677 (2020-03-04)
In Barth syndrome (BTHS) mutations in tafazzin leads to changes in both the quantities and the molecular species of cardiolipin (CL), which are the hallmarks of BTHS. Contrary to the well-established alterations in CL associated with BTHS; recently a marked
Akira Abe et al.
Journal of lipid research, 48(10), 2255-2263 (2007-07-14)
A novel lysosomal phospholipase A(2) (LPLA2) with specificity toward phosphatidylethanolamine and phosphatidylcholine was previously purified and cloned. LPLA2 transfers sn-1 or sn-2 acyl groups of phospholipids to the C1 hydroxyl of the short-chain ceramide N-acetylsphingosine (NAS) under acidic conditions. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service