Skip to Content
Merck
All Photos(1)

Key Documents

870122P

Avanti

18:1 Caproylamine PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(hexanoylamine)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€152.00

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€152.00

About This Item

Empirical Formula (Hill Notation):
C47H89N2O9P
CAS Number:
Molecular Weight:
857.19
UNSPSC Code:
12352211
NACRES:
NA.25

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870122P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN(C(CCCCC[NH3+])=O)[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C47H89N2O9P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33-37-46(51)55-42-44(43-57-59(53,54)56-41-40-49-45(50)36-32-31-35-39-48)58-47(52)38-34-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,44H,3-16,21-43,48H2,1-2H3,(H,49,50)(H,53,54)/b19-17-,20-18-/t44-/m1/s1

InChI key

OVCGTEBXOCFVEO-OHRCZFALSA-N

General description

18:1 Caproylamine PE or (1,2-Dioleoyl-sn-Glycero-3-Phosphoethanolamine- N-(hexanoylamine), belongs to a group of head group modified functionalized lipids. Functionalized lipids are commonly used as reporting molecules.[1]

Application

18:1 Caproylamine PE may be used as an indicator in competitive enzyme-linked immunosorbent assay (ELISA) for determining affinities between glycolipid analogs and human or mouse CD1d molecules.[2] It may also be used as a lipid antigen for CD1d-Ig–based artificial antigen-presenting cells (aAPC) production.[3]

Packaging

5 mL Clear Glass Sealed Ampule (870122P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangming Li et al.
Journal of immunology (Baltimore, Md. : 1950), 183(7), 4415-4421 (2009-09-08)
C-glycoside analogues of alpha-galactosylceramide were shown to activate both human and mouse invariant NKT (iNKT) cells. Among these analogues, GCK152, which has an aromatic ring in the acyl chain, exhibited a stronger stimulatory activity against human iNKT cells and a
Tonya J Webb et al.
Cancer research, 72(15), 3744-3752 (2012-06-01)
Tumors often display mechanisms to avoid or suppress immune recognition. One such mechanism is the shedding of gangliosides into the local tumor microenvironment, and a high concentration of circulating gangliosides is associated with poor prognosis. In this study, we identify
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service