Skip to Content
Merck
All Photos(1)

Key Documents

880270P

Avanti

C8 PEG5000 Ceramide

Avanti Research - A Croda Brand 880270P, powder

Synonym(s):

N-octanoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)5000]}

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€458.00

€458.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€458.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€458.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880270P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880270P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC)=O)COC(CCC([(OCH2CH2)113OCH3])=O)=O

General description

Ceramides are heterogeneous lipid molecules, prominent in the lamellar sheets of stratum corneum. They belong to the group of sphingolipids.[1]

Biochem/physiol Actions

The polymer size and acyl chain length of poly(ethylene glycol) (PEG) conjugated ceramides are known to influence the transfection potency of stabilized plasmid-lipid particle (SPLP) and its formation.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (880270P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stabilized plasmid-lipid particles: factors influencing plasmid entrapment and transfection properties
Mok KWC, et al.
Biochimica et Biophysica Acta - Biomembranes, 1419(2), 137-150 (1999)
Ceramides and skin function
Coderch L, et al.
American Journal of Clinical Dermatology, 4(2), 107-129 (2003)
Hudson P Pace et al.
Analytical chemistry, 90(21), 13065-13072 (2018-10-24)
Over the last two decades, supported lipid bilayers (SLBs) have been extensively used as model systems to study cell membrane structure and function. While SLBs have been traditionally produced from simple lipid mixtures, there has been a recent surge in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service