Skip to Content
Merck
All Photos(1)

Key Documents

855725C

Avanti

2-18:1 Lyso PE

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1-Hydroxy-2-oleoyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€404.00

€404.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€404.00

About This Item

Empirical Formula (Hill Notation):
C23H46NO7P
CAS Number:
Molecular Weight:
479.59
UNSPSC Code:
51191904
NACRES:
NA.25

€404.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL (855725C-10MG)
pkg of 1 × 1 mL (855725C-1MG)
pkg of 1 × 1 mL (855725C-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL in chloroform (855725C-1MG)
10 mg/mL in chloroform (855725C-10MG)
5 mg/mL in chloroform (855725C-5MG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[NH3+])(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)CO)=O

InChI

1S/C23H46NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(26)31-22(20-25)21-30-32(27,28)29-19-18-24/h9-10,22,25H,2-8,11-21,24H2,1H3,(H,27,28)/b10-9-/t22-/m1/s1

InChI key

NOVZJYYJYIFXJC-MZMPXXGTSA-N

General description

2-18:1 Lyso PE is a lyso phosphatidylethanolamine with oleic acid substitution at sn-2 position.[1] LPE is synthesized from phosphatidylethanolamine (PE) through the action of phospholipase A.[2]

Application

2-18:1 Lyso PE is suitable for use as standard, as sn-2 isomers of lyso PE for hydrophilic interaction liquid chromatography-paired with ion mobility-mass spectrometry (HILIC-IM-MS),[1] and high performance thin-layer chromatography (HPTLC).[3]

Biochem/physiol Actions

Lyso phosphatidylethanolamine (Lyso PE) is a lipid present naturally in plants and it retards senescence in leaves, postharvest fruits and flowers.[4] LPE has antifungal and antibacterial activity. LPE functions to induce neuronal differentiation, increases intracellular calcium, stimulates chemotactic migration and cellular invasion in cancer cells.[2]

Packaging

5 mL Clear Glass Sealed Ampule (855725C-1MG, 855725C-5MG, and 855725C-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipidomic consequences of phospholipid synthesis defects in Escherichia coli revealed by HILIC-ion mobility-mass spectrometry.
Hines KM and Xu L
Chemistry and Physics of Lipids, 219, 15-22 (2019)
Trypanosoma cruzi epimastigotes store cholesteryl esters in lipid droplets after cholesterol endocytosis.
Pereira MG, et al.
Molecular and Biochemical Parasitology, 224, 6-16 (2018)
Emerging lysophospholipid mediators, lysophosphatidylserine, lysophosphatidylthreonine, lysophosphatidylethanolamine and lysophosphatidylglycerol.
Makide K, et al.
Prostaglandins & other lipid mediators, 89(3-4), 135-139 (2009)
Inhibition of phospholipase D by lysophosphatidylethanolamine, a lipid-derived senescence retardant.
Ryu SB, et al.
Proceedings of the National Academy of Sciences of the USA, 94(23), 12717-12721 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service