Skip to Content
Merck
All Photos(2)

Key Documents

846725C

Avanti

18:1 Lyso PE

1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1--(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine; PE(18:1(9Z)/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€209.00

€209.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€209.00

About This Item

Empirical Formula (Hill Notation):
C23H46NO7P
CAS Number:
Molecular Weight:
479.59
UNSPSC Code:
51191904
NACRES:
NA.25

€209.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (846725C-25mg)
pkg of 2 × 20 mL (846725C-1g)
pkg of 2 × 4 mL (846725C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 846725C

concentration

10 mg/mL (846725C-25mg)
25 mg/mL (846725C-1g)
25 mg/mL (846725C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI key

PYVRVRFVLRNJLY-MZMPXXGTSA-N

General description

Lyso-phosphatidylethanolamine (lysoPE) is a structural isomer which carries an oleic acid at the sn-1 position.[1] This lipid occurs naturally.[2]

Application

18:1 Lyso PE (1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine) has been used as a lipid standard in the optimization of the liquid chromatography−mass spectrometry (LC-MS) method conditions.[3] It may be used to study its effects on [Ca2+]i in PC-12 neuronal cells,[4] MDA-MB-231 and SK-OV3 cells.[5]

Biochem/physiol Actions

Lyso-phosphatidylethanolamine (lysoPE) can serve as a substrate for SocA, the short-chain alcohol dehydrogenase.[6] It helps to delay the senescence in the plant parts.[2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (846725C-1g)
5 mL Clear Glass Sealed Ampule (846725C-200mg)
5 mL Clear Glass Sealed Ampule (846725C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly M Hines et al.
Chemistry and physics of lipids, 219, 15-22 (2019-01-21)
Our understanding of phospholipid biosynthesis in Gram-positive and Gram-negative bacteria is derived from the prototypical Gram-negative organism Escherichia coli. The inner and outer membranes of E. coli are largely composed of phosphatidylethanolamine (PE), minor amounts of phosphatidylglycerol (PG) and cardiolipin
Soo-Jin Park et al.
Biomolecules & therapeutics, 22(2), 129-135 (2014-04-23)
Previously, we reported that lysophosphatidylethanolamine (LPE), a lyso-type metabolite of phosphatidylethanolamine, can increase intracellular Ca(2+) ([Ca(2+)]i) via type 1 lysophosphatidic acid (LPA) receptor (LPA1) and CD97, an adhesion G-protein-coupled receptor (GPCR), in MDA-MB-231 breast cancer cells. Furthermore, LPE signaling was
S B Ryu et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(23), 12717-12721 (2000-10-20)
Phospholipid signaling mediated by lipid-derived second messengers or biologically active lipids is still new and is not well established in plants. We recently have found that lysophosphatidylethanolamine (LPE), a naturally occurring lipid, retards senescence of leaves, flowers, and postharvest fruits.
Madhavi Avadhani et al.
Journal of bacteriology, 188(24), 8543-8550 (2006-10-10)
Short-chain alcohol dehydrogenases (SCADHs) synthesize a variety of intercellular signals and other chemically diverse products. It is difficult to predict the substrate of a SCADH on the basis of amino acid sequence homology, as the substrates are not known for
Jung-Min Lee et al.
Biochemical and biophysical research communications, 461(2), 378-382 (2015-04-19)
G protein-coupled receptors (GPCRs) have been implicated in lysophosphatidylethanolamine (LPE)-induced increases in intracellular Ca(2+) ([Ca(2+)]i), but in different cell types, this response may be dependent or independent of lysophosphatidic acid (LPA) GPCR. The effects of LPEs from Grifola frondosa on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service