Skip to Content
Merck
All Photos(2)

Key Documents

855476P

Avanti

13:0 Lyso PC

1-tridecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-tridecanoyl-sn-glycero-3-phosphocholine; PC(13:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€163.00

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C21H44NO7P
CAS Number:
Molecular Weight:
453.55
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855476P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855476P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCC)=O

InChI

1S/C21H44NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-21(24)27-18-20(23)19-29-30(25,26)28-17-16-22(2,3)4/h20,23H,5-19H2,1-4H3/t20-/m1/s1

InChI key

WNRCJJWBAXNAPE-HXUWFJFHSA-N

Application

13:0 Lyso PC has been used:
  • as an internal standard: for the quantification of targeted infant metabolites[1]
  • used as an internal standard in liquid chromatography-mass spectrometry (LC-MS) to identify changes in lipid profile of eggs[2]
  • as an internal standard for tandem mass spectrometry analysis of lipid species[3]

Packaging

5 mL Amber Glass Screw Cap Vial (855476P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipidomic investigation of eggs' yolk: Changes in lipid profile of eggs from different conditions
Campos AM, et al.
Food Research International, 89, 177-185 (2016)
The impact of human breast milk components on the infant metabolism
Hellmuth C, et al.
PLoS ONE, 13(6) (2018)
The impact of human breast milk components on the infant metabolism
Hellmuth C, et al.
Testing, 13(6) (2018)
High-Throughput Plasma Lipidomics: Detailed Mapping of the Associations with Cardiometabolic Risk Factors
Huynh K, et al.
Cell Chemical Biology, 26(1), 71-84 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service