Skip to Content
Merck
All Photos(2)

Key Documents

840503P

Avanti

18:0-18:1 PG

1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

1-octadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); SOPG; PG(18:0/18:1(9Z)); 110652

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€143.00
500 MG
€772.00

€143.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€143.00
500 MG
€772.00

About This Item

Empirical Formula (Hill Notation):
C42H80O10PNa
CAS Number:
Molecular Weight:
799.04
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€143.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 5 × 100 mg (840503P-500mg)
pkg of 1 × 25 mg (840503P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840503P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H81O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,39-40,43-44H,3-17,19,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b20-18-;/t39?,40-;/m1./s1

InChI key

AZUBILBUHSANJX-VDVGXYCESA-M

General description

18:0-18:1 PG (1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) is a phospholipid which carries a small head group.[1] It is made of two 18 carbon chains with phosphoglycerol backbone.

Application

18:0-18:1 PG (1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) has been used as a standard to analyze total lipid extracts by thin-layer chromatography (TLC).[2] It may be used to measure the T-cell responses to lipid antigens using CD1 antigen-presenting molecule (CD1b) tetramers[1] and in liposomes preparation.[3]

Biochem/physiol Actions

Phosphatidylglycerol (PG) participates in photosynthetic electron transport.[4]

Packaging

5 mL Clear Glass Sealed Ampule (840503P-25mg)
5 mL Clear Glass Sealed Ampule (840503P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Reinink et al.
The Journal of experimental medicine, 216(4), 757-771 (2019-02-26)
Salmonella species are among the world's most prevalent pathogens. Because the cell wall interfaces with the host, we designed a lipidomics approach to reveal pathogen-specific cell wall compounds. Among the molecules differentially expressed between Salmonella Paratyphi and S. Typhi, we
Koichi Kobayashi et al.
Plant cell reports, 34(4), 631-642 (2014-12-06)
With phosphate deficiency, the role of phosphatidylglycerol is compensated by increased glycolipid content in thylakoid membrane biogenesis but not photosynthetic electron transport in Arabidopsis chloroplasts. In plants and cyanobacteria, anionic phosphatidylglycerol (PG) is the only major phospholipid in thylakoid membranes
J P Michel et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2967-2979 (2015-09-08)
Bacterial resistance to antibiotics has become today a major public health issue. In the development of new anti-infectious therapies, antimicrobial peptides appear as promising candidates. However, their mechanisms of action against bacterial membranes are still poorly understood. We describe for
Adam Shahine et al.
Nature communications, 10(1), 56-56 (2019-01-06)
CD1 proteins are expressed on dendritic cells, where they display lipid antigens to T-cell receptors (TCRs). Here we describe T-cell autoreactivity towards ubiquitous human membrane phospholipids presented by CD1b. These T-cells discriminate between two major types of lipids, sphingolipids and
Joury S van 't Klooster et al.
eLife, 9 (2020-04-18)
Yeast tolerates a low pH and high solvent concentrations. The permeability of the plasma membrane (PM) for small molecules is low and lateral diffusion of proteins is slow. These findings suggest a high degree of lipid order, which raises the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service