Skip to Content
Merck
All Photos(2)

Key Documents

810890C

Avanti

Fluorescent DOTAP

1-oleoyl-2-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-3-trimethylammonium propane (chloride salt), chloroform

Synonym(s):

18:1-06:0 NBD DOTAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60ClN5O7
CAS Number:
Molecular Weight:
710.34
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810890C-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810890C

concentration

0.05 mg/mL (810890C-50ug)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCNC1=CC=C([N+]([O-])=O)C2=NON=C12)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Cl-]

General description

N-(1-(2,3-dioleoyloxy)propyl)-N,N,N-trimethylammonium (DOTAP) is a cationic lipid, which acts as a transfection agent. It forms strong complexes with DNA and aids the cellular delivery of RNA and DNA.

Packaging

5 mL Clear Glass Sealed Ampule (810890C-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurons: Methods and Applications for the Cell Biologist (2003)
Phospholipid-cationic lipid interactions: influences on membrane and vesicle properties
Campbell RB, et al.
Biochimica et Biophysica Acta - Biomembranes, 1512(1), 27-39 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service