WHA230300
Whatman® drain disc
diam. 10 mm
Synonym(s):
Whatman filter, Whatman filters, Whatman paper, Z741469
Select a Size
€70.20
Select a Size
About This Item
€70.20
Recommended Products
material
polyethylene housing
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 230300
Whatman Article No. 28418857 (US reference)
diam.
10 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Applications
- General laboratory microfiltration
- Quality control and sterility testing
- Removal of particulates from HPLC solvents
- Tissue culture media filtration
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service