Skip to Content
Merck
All Photos(1)

Documents

610034

Avanti

Syringe Needle

Avanti Research - A Croda Brand

Synonym(s):

Extruder Syringe Needle, Replacement Needle for Syringe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NB.03

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Replacement Part for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research Mini-Extruder.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service