610025
Avanti
Membrane Support-PTFE
Avanti Research™ - A Croda Brand
Synonym(s):
Membrane Support-PTFE, PTFE Internal Membrane Supports with O-rings (2)
Select a Size
About This Item
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
15-25°C
General description
Legal Information
also commonly purchased with this product
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service