600511
Avanti
5mL Amber SCHOTT® FIOLAX® Vial with PTFE-lined Screw Cap Closure (3 pack)
Avanti Research™ - A Croda Brand
Synonym(s):
5mL Amber SCHOTT® FIOLAX® Vial with PTFE-lined Screw Cap Closure (3 pack), 5mL Amber SCHOTT FIOLAX Vial with PTFE-lined Screw Cap Closure
Select a Size
€17.90
Select a Size
About This Item
€17.90
Recommended Products
description
5mL Amber SCHOTT FIOLAX Vial with PTFE-lined Screw Cap Closure (3 pack)
packaging
vial of 3 × 5 mL (600511-1EA)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
ambient
storage temp.
room temp
Related Categories
Packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service