Skip to Content
Merck
All Photos(1)

Key Documents

27151

Supelco

Vials, screw top, clear glass (vial only)

volume 7 mL, clear glass vial, thread for 15-425, pkg of 100 ea

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€68.60

€68.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 EA
€68.60

About This Item

UNSPSC Code:
41122001

€68.60


Please contact Customer Service for Availability

Request a Bulk Order

material

clear glass vial

feature

closure type screw top vial

packaging

pkg of 100 ea

O.D. × H × I.D.

17 mm × 60 mm × 9.5 mm

volume

7 mL

fitting

thread for 15-425

Looking for similar products? Visit Product Comparison Guide

General description

Vials are suitable basically to inject samples from an autosampler. They are available in various sizes, although 2 mL vials are more commonly used. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials.[1]

Application

It has been used in vapour trap for determining atmospheric water vapour for isotopic analysis.[2] It has also been used in solid phase microextraction (SPME) combined with gas chromatography/mass spectrometry (GC/MS) method during the determination of oxidation of freeze-dried chicken myofibrils spiked with methyl linoleate.[3]

Storage Class Code

NOHAZ - NOHAZ : Non Hazardous Substance


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oona McPolin. Taylor
An Introduction to HPLC for Pharmaceutical Analysis, 49-49 (2009)
Solid phase microextraction- gas chromatography for quantifying headspace hexanal above freeze-dried chicken myofibrils.
Goodridge, Carolyn F., et al.
Journal of Agricultural and Food Chemistry, 51.15, 4185-4190 (2003)
A rapid method for the sampling of atmospheric water vapour for isotopic analysis.
Peters, Leon I., and Dan Yakir.
Rapid Communications in Mass Spectrometry, 24.1, 103-108 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service