Key Documents
Z742587
Accuris M1 Grade Calibration Weight
size 2000 g, stainless steel
Select a Size
1 EA
€197.00
€197.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€197.00
About This Item
UNSPSC Code:
41111500
NACRES:
NB.71
€197.00
Please contact Customer Service for Availability
material
stainless steel
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific W1005-2000
size
2000 g
General description
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)