Key Documents
Z741340
MAGic Clamp universal platform for flasks and tube racks
L × W 16 in. × 16.5 in.
Select a Size
1 EA
€445.00
€445.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€445.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.77
€445.00
Please contact Customer Service for Availability
manufacturer/tradename
Benchmark Scientific BT4010-MR
L × W
16 in. × 16.5 in.
General description
The convenient MAGic Clamp platform can be used for both Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
Legal Information
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)