Key Documents
Z565008
Vacuum accessories for Buchi® vacuum pumps
Valve unit
Synonym(s):
Vacuum Pump Accessories
Select a Size
1 EA
€848.00
€848.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€848.00
About This Item
UNSPSC Code:
41105101
NACRES:
NB.43
€848.00
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
Büchi 47160
General description
This valve comes with a Woulff bottle and an attachment for Rotavapors. Not to be used with V700/710 pump in stand alone systems.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)