Z693154
Norell® Sample Vault Series™ NMR tubes
standard wall, open cap, frequency 700 MHz, diam. × L 5 mm × 178 mm
Synonym(s):
NMR tube for 96 position carriers, NMR tubes for lab automation systems
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
colorless tube
feature
neck thread push fit
packaging
pkg of 96 ea
manufacturer/tradename
Norell SVOP-5-178-96PK
parameter
700 MHz frequency
camber
60 μm
diam. × L
5 mm × 178 mm
wall thickness
0.38 mm
Looking for similar products? Visit Product Comparison Guide
General description
For Bruker® SampleJet™ robotic high-throughput automated systems. NMR tubes with NorLoc™ caps are designed to be used with 96 position carriers using 103.5 mm (4 in.) long NMR tubes with open port caps, or 178 mm (7 in.) long NMR tubes with closed port caps. NorLoc caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument.
Patent pending design incorporates a blue band positioned on the NMR tube that aligns with the base of the cap, indicating a properly locked position. The NorLoc cap and Sample Vault Series tube is an integrated closure system for fail-safe delivery of your sample to the magnet. Tubes are available in both 3 mm and 5 mm diameters for use up to 700 MHz and up to 950 MHz.
Patent pending design incorporates a blue band positioned on the NMR tube that aligns with the base of the cap, indicating a properly locked position. The NorLoc cap and Sample Vault Series tube is an integrated closure system for fail-safe delivery of your sample to the magnet. Tubes are available in both 3 mm and 5 mm diameters for use up to 700 MHz and up to 950 MHz.
- Engineered for new generation high-throughput lab automation systems
- Two precision types for up to 700 MHz and up to 950 MHz spectrometers
- Superior NorLoc cap attaches semi-permanently for multiple use and for critical applications
- Choice of either NorLoc closed port cap or open port cap
- Closed port caps are supplied with tubes in a bag, open port caps are installed on the tubes
- Safe for cold refrigeration storage, works with cryo-probes and variable temperature studies
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
NorLoc is a trademark of Norell, Inc.
Norell is a registered trademark of Norell, Inc.
Sample Vault Series is a trademark of Norell, Inc.
SampleJet is a trademark of Bruker-Physik AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service