Skip to Content
Merck
All Photos(1)

Key Documents

Z566845

Plas-Labs PCR UV chamber germicidal light package

rated at 254 nm

Synonym(s):

isolation chambers, pcr chambers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€940.00

€940.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€940.00

About This Item

UNSPSC Code:
41103406
NACRES:
NB.35

€940.00


Please contact Customer Service for Availability

manufacturer/tradename

Plas-Labs 800-UV/18

technique(s)

PCR: suitable

Related Categories

General description

The PCR UV germicidal light package is rated at 254 nm and will decontaminate all surfaces of the interior of the Plas-Labs PCR UV chamber. The lamp should be activated 15 minutes prior to and 15 minutes after planned use.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service