Skip to Content
Merck
All Photos(1)

Key Documents

Z412163

Hermetically sealed oil mist eliminator

For Welch Models 1400N, 1402N, and 1376N

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,210.00

€1,210.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€1,210.00

About This Item

UNSPSC Code:
41105101
NACRES:
NB.79

€1,210.00


Please contact Customer Service for Availability

feature

For Welch Models 1400N, 1402N, and 1376N

manufacturer/tradename

Welch


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service