Key Documents
Z177288
Sigma-Aldrich
Stainless steel clamp
Overall L 8 1/2 in.
Select a Size
1 EA
€98.50
€98.50
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€98.50
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
€98.50
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18354-0000
Overall L
8 1/2 in.
General description
Tri-grip, 0 to 1 in. opening range. Has spring-action opening with flat wing-head screw for clamping. Jaws covered with rubber tubing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)