Skip to Content
Merck
All Photos(2)

Key Documents

Z162299

Sigma-Aldrich

Aldrich® solid-phase peptide synthesis flask

capacity 1 L, center joint: ST/NS 24/40

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€805.00

€805.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€805.00

About This Item

UNSPSC Code:
41121804
NACRES:
NB.24

€805.00


Please contact Customer Service for Availability

material

borosilicate glass
four-neck
round bottom

Joint

center joint: ST/NS24/40

sterility

non-sterile

feature

closure type Standard Taper Joint

packaging

pack of 1

manufacturer/tradename

Sigma-Aldrich

capacity

1 L

Looking for similar products? Visit Product Comparison Guide

General description

Has three 24/40 side joints and a center joint that varies in size according to the capacity of the flask. Coarse frit (25 to 50 μm) and PTFE Rotaflo® stopcock permit rapid filtration and resin washing.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Rotaflo is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service