Skip to Content
Merck
All Photos(1)

Key Documents

Z146242

Sigma-Aldrich

Scienceware® plastic-tubing cutter, replacement blade

size 7/8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€19.90

€19.90


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€19.90

About This Item

UNSPSC Code:
41122406
NACRES:
NB.35

€19.90


Please contact Customer Service for Availability

manufacturer/tradename

Bel-Art

size

7/8 in.

Related Categories

General description

Portable, pocket-sized cutter cuts flexible tubing from 1.6mm to 19mm (¹⁄₁₆ to ¾") O.D.

Features and Benefits

  • Spring-loaded jaw will remain closed for safety when not in use
  • Replaceable, hardened steel blade makes clean, straight cuts

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service