Key Documents
Z118990
Sigma-Aldrich
Scienceware® Frigimat® Junior dry ice maker
produces dry ice in snowflake form
Select a Size
1 EA
€289.00
€289.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€289.00
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
€289.00
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F38886-0000
General description
Produces dry ice in snowflake form. Consists of a special brass nozzle, PE cone, and a 6 × 12 in. heavy-duty cloth bag collector. The nozzle is attached directly to a cylinder (must have dip tube) of liquid CO2.
Ideal for use in remote locations, low demand or for packing or freezing samples. All components are included to connect to a standard CO2 cylinder with siphon tube to make dry ice as needed. Fittings and collection bag attach directly to the CO2 cylinder. Fits US connections.
Easy and fast dry ice flake or snow production on demand.
This device connects to CO2 gas cylinders with CGA 320 threads which is the standard for CO2 in the USA.
The Frigimate Junior Dry-Ice Maker does not fit European Gas-bottles.
Legal Information
Frigimat is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)