Skip to Content
Merck
All Photos(1)

Documents

Z558567

Sigma-Aldrich

Aldrich® microscale glassware kit

Deluxe

Synonym(s):

Glassware kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Components:
Z684139 Adapter, Claisen, 14/10 (1)
Z684163 Adapter, Thermometer, 14/10 (1)
Z684155 Adapter, Multipurpose, 14/10 (1)
Z684147 Adapter, Distilling, 14/10 (1)
Z684171 Adapter, Vacuum, 105D, 14/10 (1)
Z684236 Condenser, Air Reflux, 14/10 (1)
Z684244 Condenser, Jacketed Reflux, 14/10 (1)
Z684252 Distillation Head, Hickman w/sideport (1)
Z684260 Flask, Round Bottom, 10 mL, 14/10 (1)
Z684279 Flask, Round Bottom, 25 mL, 14/10 (1)
Z684287 Spin Bar PTFE, 2 × 7 mm (1)
Z684295 Spin Vane, 3 & 5 mL (1)
Z684309 Syringe, Glass, 1 mL (1)
Z684341 Tube, Craig, Complete, 2 mL (1)
Z684368 Tube, Drying, 14/10 (1)
Z684384 Tube, GC Collection, 5/5 (2)
Z684392 Tube, Sublimation (1)
Z684406 Vial, Conical, 0.1 mL (2)
Z684422 Vial, Conical, 3.0 mL (1)
Z684430 Vial, Conical, 5.0 mL (2)
Z684449 Vial, Conical, Thin Wall, 5 mL (1)
Z684198 Kit Box (1)
Z684228 Kit Foam (1)
High-quality, low cost glassware kits designed to cover a broad range of organic reactions in student labs and chemical research. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory. 14/10 ground-glass joints with holed-caps and O-ring seals provide secure, gas-tight connections.
Economy kit - supplied in a compartmentalized PP storage box with hinged lid. Will do 95% of experiments described in the textbook.
Deluxe kit - supplied in a foam-lined plastic carrying case. Covers all experiments in the textbook.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service