Skip to Content
Merck
All Photos(1)

Key Documents

W526401

Sigma-Aldrich

6-Acetyl-1,1,2,4,4,7-hexamethyltetralin

≥98%

Synonym(s):

1,2,3,4-Tetrahydro-1,1,3,4,4,6-hexamethyl-7-naphthyl methyl ketone, 1-(5,6,7,8-Tetrahydro-3,5,5,6,8,8-hexamethyl-2-naphthalenyl)ethanone, 5′,6′,7′,8′-Tetrahydro-3′,5′,5′,6′,8′,8′-hexamethyl-2′-acetonaphthone, 6-Acetyl-1,1,2,4,4,7-hexamethyl-1,2,3,4-tetrahydronaphthalene, AHMT, Fixolide, Musk tonalid, NSC 19550

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O
CAS Number:
Molecular Weight:
258.40
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
Fragrance grade
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥98%

mp

52-57 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

ethereal

SMILES string

CC1CC(C)(C)c2cc(C(C)=O)c(C)cc2C1(C)C

InChI

1S/C18H26O/c1-11-8-16-15(9-14(11)13(3)19)17(4,5)10-12(2)18(16,6)7/h8-9,12H,10H2,1-7H3

InChI key

DNRJTBAOUJJKDY-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Avila et al.
Chemosphere, 81(9), 1137-1142 (2010-09-25)
A continuous injection experiment was implemented in a pilot-scale horizontal subsurface flow constructed wetland system to evaluate the behavior of four pharmaceuticals and personal care products (i.e. ibuprofen, naproxen, diclofenac and tonalide) and a phenolic estrogenic compound (i.e. bisphenol A).
Zhengjun Hu et al.
Environmental toxicology and chemistry, 29(9), 1877-1882 (2010-09-08)
We measured two nitro musk fragrances (musk xylene) and musk ketone) and five polycyclic musk fragrances (galaxolide [HHCB], tonalide [AHTN], celestolide [ADBI], traseolide [ATII], and phantolide [AHMI]) in human blood samples from 11 cities of China (n = 204). Possible
Yu-Jie He et al.
The Science of the total environment, 447, 248-254 (2013-02-07)
The presence and distribution of typical of pharmaceuticals and personal care products (PPCPs), which comprise two types of polycyclic musks (PCMs) including Galaxolide (HHCB) and Tonalide (AHTN) as well as six types of estrogens containing estrone (E1), 17β-estradiol (E2), estriol
Zhengjun Hu et al.
Chemosphere, 84(11), 1630-1635 (2011-06-01)
Seven typical synthetic musks (SMs) in the samples from the surface water, sediment and fish of the Haihe River were measured. The SM concentrations in the sediment and surface water of the Haihe River were significantly lower than those in
Javier Santiago-Morales et al.
Water research, 46(14), 4435-4447 (2012-06-20)
Synthetic musks have been reported in wastewaters at concentrations as high as tens of micrograms per litre. The two most significant polycyclic musk fragrance compounds are 1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta(g)-2-benzopyran (HHCB, trade name galaxolide®) and 7-acetyl-1,1,3,4,4,6-hexamethyltetrahydronaphthalene (AHTN, trade name tonalide®). We report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service