Skip to Content
Merck
All Photos(1)

Key Documents

W353108

Sigma-Aldrich

Cyclohexanecarboxylic acid

≥98%, FG

Synonym(s):

Hexahydrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CO2H
CAS Number:
Molecular Weight:
128.17
FEMA Number:
3531
Beilstein:
970529
EC Number:
Council of Europe no.:
11911
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.060
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥98%

refractive index

n20/D 1.461 (lit.)

bp

232-233 °C (lit.)

mp

29-31 °C (lit.)

density

1.033 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; acidic

SMILES string

OC(=O)C1CCCCC1

InChI

1S/C7H12O2/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2,(H,8,9)

InChI key

NZNMSOFKMUBTKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanecarboxylic acid occurs naturally in the leaves of Ormosia henryi Prain. It is also one of the aromatic volatile compounds found in the secretion of tail glands of red deer.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new sexual signal in rutting male red deer: Age related chemical scent constituents in the belly black spot.
Martin J, et al.
Mammalian Biology-Zeitschrift fur Saugetierkunde, 79(6), 362-368 (2014)
GC-MS Analysis of Chemical Constituents of the Volatile Oil from Leaves of Ormosia henryi Prain.
Jiexiong NBZWF & Mi T.
Guangdong Forestry Science and Technology, 2, 012-012 (2012)
Housna Mouttaki et al.
Environmental microbiology, 10(12), 3265-3274 (2008-08-19)
In methanogenic environments, the main fate of benzoate is its oxidization to acetate, H(2) and CO(2) by syntrophic associations of hydrogen-producing benzoate degraders and hydrogen-using methanogens. Here, we report the use of benzoate as an electron acceptor. Pure cultures of
Christo Christov et al.
ChemMedChem, 6(1), 131-140 (2010-12-07)
2-(3,5-Dichlorophenylcarbamoyl)cyclohexanecarboxylic acid (1) is a potent and selective positive allosteric modulator of metabotropic glutamate receptor subtype 4 (mGluR4). The activity of 1 was reported to reside in the cis diastereomer with equal potency between its enantiomeric forms (Niswender et al., Mol. Pharmacol.
P G Egland et al.
Journal of bacteriology, 177(22), 6545-6551 (1995-11-01)
The first step of anaerobic benzoate degradation is the formation of benzoyl-coenzyme A by benzoate-coenzyme A ligase. This enzyme, purified from Rhodopseudomonas palustris, is maximally active with 5 microM benzoate. To study the molecular basis for this reaction, the benzoate-coenzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service