Skip to Content
Merck
All Photos(1)

Key Documents

W317410

Sigma-Aldrich

4-Hydroxy-2,5-dimethyl-3(2H)-furanone

15 wt. % (in propylene glycol), FG

Synonym(s):

2,5-Dimethyl-4-hydroxy-3(2H)-furanone, Furaneol, Strawberry furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
FEMA Number:
3174
Beilstein:
1281357
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.010
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

concentration

15 wt. % (in propylene glycol)

refractive index

n20/D 1.444

mp

73-77 °C (lit.)

density

1.057 at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; strawberry; sweet

SMILES string

CC1OC(C)=C(O)C1=O

InChI

1S/C6H8O3/c1-3-5(7)6(8)4(2)9-3/h3,8H,1-2H3

InChI key

INAXVXBDKKUCGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

210.2 °F

Flash Point(C)

99 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silke Illmann et al.
Journal of agricultural and food chemistry, 57(7), 2889-2895 (2009-03-05)
The formation of 4-hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF) was studied in aqueous model systems containing L-rhamnose and L-lysine. The approach consisted in systematically varying four reaction parameters (rhamnose concentration, rhamnose to lysine ratio, pH, and phosphate concentration) at 3 levels. A fractional factorial
Fernando de la Peña Moreno et al.
Journal of chromatography. A, 1217(7), 1083-1088 (2009-11-03)
A method based on the use of the through oven transfer adsorption-desorption (TOTAD) interface in on-line coupling between reversed phase liquid chromatography and gas chromatography (RPLC-GC) for the determination of chiral volatile compounds was developed. In particular, the method was
Thomas Raab et al.
The Plant cell, 18(4), 1023-1037 (2006-03-07)
The flavor of strawberry (Fragaria x ananassa) fruit is dominated by an uncommon group of aroma compounds with a 2,5-dimethyl-3(H)-furanone structure. We report the characterization of an enzyme involved in the biosynthesis of 4-hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF; Furaneol), the key flavor compound
Goreti Botelho et al.
Analytica chimica acta, 657(2), 198-203 (2009-12-17)
To evaluate the potential aroma of Aragonez clonal red musts, several free and glycosidically bound odourant compounds were extracted. Then, the gas chromatography-olfactometry (GC-O) posterior intensity method was used to evaluate their odour intensity and the compounds were identified by
Nicole Christina Stadler et al.
Journal of agricultural and food chemistry, 57(9), 3949-3954 (2009-04-03)
A number of 3(2H)-furanones are synthesized by fruits and have been found in cooked foodstuffs, where they impart flavor and odor because of their low perception thresholds. They show genotoxic properties in model studies but are also ranked among the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service