Skip to Content
Merck
All Photos(1)

Key Documents

W216900

Sigma-Aldrich

Buchu leaf oil, Crenulata

natural, FG

Synonym(s):

Agathosma crenulata

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2169
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Kosher
natural
biological source:
Barosma crenulata
food allergen:
no known allergens

biological source

Barosma crenulata

Quality Level

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.510

form

liquid

composition

Limonene, 15.7%
Linalool, 0.6%
Pulegone, 15-23%

refractive index

n20/D 1.476

density

0.915 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; green; fruity; tropical; minty; herbaceous

Physical properties

black current like odor

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service