Key Documents
TEM-CF300CU50
Sigma-Aldrich
Carbon Film Supported Copper Grid
Standard Thickness, size 300 mesh, box of 50 ×
Synonym(s):
Carbon Film 300 Mesh, Copper
About This Item
UNSPSC Code:
41111761
material
carbon film
copper grid
packaging
box of 50 ×
film size
5-6 nm
size
300 mesh
General description
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)