Key Documents
930369
Sigma-Aldrich
TEM Grids
Carbon Film Supported Copper Grid, Standard Thickness, grid size 200 mesh, box of 50
Synonym(s):
TEM grids
Select a Size
1 EACH
€130.00
€130.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€130.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
€130.00
Please contact Customer Service for Availability
Product Name
Carbon Film Supported Copper Grid, Standard Thickness, grid size 200 mesh, box of 50
material
carbon film
copper grid
packaging
box of 50
film size
5-6 nm
grid size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported standard square mesh grids.
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)