T61107
Triethyl phosphate
99%
Synonym(s):
Ethyl phosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5O)3PO
CAS Number:
Molecular Weight:
182.15
Beilstein:
1705772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
vapor density
6.28 (vs air)
vapor pressure
1 mmHg ( 40 °C)
Assay
99%
autoignition temp.
845 °F
expl. lim.
10 %
refractive index
n20/D 1.403 (lit.)
bp
215 °C (lit.)
density
1.072 g/mL at 25 °C (lit.)
SMILES string
CCOP(=O)(OCC)OCC
InChI
1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3
InChI key
DQWPFSLDHJDLRL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
240.8 °F - closed cup
Flash Point(C)
116 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shujuan Meng et al.
Water research, 83, 248-257 (2015-07-15)
Alginate, a linear copolymer, is composed of 1,4-linked β-d-mannuronic acid (M) and α-l-guluronic acid (G), which are combined into homopolymeric blocks (MM-block and GG-block) and heteropolymeric block (MG-block). It has been widely used as a model foulant in various studies
B S Fales et al.
Journal of the American Society for Mass Spectrometry, 22(10), 1862-1871 (2011-09-29)
The gas-phase structures of protonated and sodium cationized complexes of triethyl phosphate, [TEP + H](+) and [TEP + Na](+), are examined via infrared multiple photon dissociation (IRMPD) action spectroscopy using tunable IR radiation generated by a free electron laser, a
T Salthammer et al.
Indoor air, 13(1), 49-52 (2003-03-01)
Organophosphate esters, halogenated and non-halogenated, are frequently used for fire protection of building materials. With regard to toxicological profiles it is desired to avoid human exposure in the indoor environment. Moreover, some hazardous volatile organic compounds detected in indoor air
Yuze Sun et al.
The Analyst, 134(7), 1386-1391 (2009-06-30)
We demonstrated rapid 2,4-dinitrotoluene (DNT) vapor detection at room temperature based on an optofluidic ring resonator (OFRR) sensor. With the unique on-column separation and detection features of OFRR vapor sensors, DNT can be identified from other interferences coexisting in the
Nele Van den Eede et al.
Environment international, 37(2), 454-461 (2010-12-24)
A new and efficient analytical method was developed and validated for the analysis of organophosphorus flame retardants (OPFRs) in indoor dust samples. This method involves an extraction step by ultrasonication and vortex, followed by extract clean-up with Florisil solid-phase extraction
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service