Skip to Content
Merck
All Photos(1)

Key Documents

8.21141

Sigma-Aldrich

Triethyl phosphate

for synthesis

Synonym(s):

Triethyl phosphate, Phosphoric acid triethyl ester, TEP

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€28.50
1 L
€88.70
10 KG
€304.00

€28.50


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€28.50
1 L
€88.70
10 KG
€304.00

About This Item

Linear Formula:
(C2H5O)3PO
CAS Number:
Molecular Weight:
182.15
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-114-5
NACRES:
NA.22

€28.50


Estimated to ship on14 April 2025Details


Request a Bulk Order

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

480 °C

potency

1165 mg/kg LD50, oral (Rat)

expl. lim.

1.2-10 % (v/v)

pH

7 (20 °C in H2O)

bp

215 °C/1013 hPa

mp

-57 °C

transition temp

flash point 130 °C

solubility

500 g/L (slow decomposition)

density

1.069 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(OCC)(OCC)OCC

InChI

1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3

InChI key

DQWPFSLDHJDLRL-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.068 - 1.069
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service