SYNF892450
Synthware™ reaction tube with glass stopcock
50 mL, joint: ST/NS 24/40
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
colorless tube
glass tube
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
graduations: no
Push fit
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F892450
capacity
50 mL
50 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With an outer joint and 2 mm glass stopcock having a constant pressure retention device.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service