Key Documents
SLW4507/45D
Pyrex® Membrane screwcap, 0.2 um PTFE membrane, GL 45 screwthread
Synonym(s):
Pyrex, SciLabware 4507/45D
Select a Size
5 EA
€45.30
€45.30
Please contact Customer Service for Availability
Select a Size
Change View
5 EA
€45.30
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€45.30
Please contact Customer Service for Availability
material
PTFE membrane
polypropylene screw cap
manufacturer/tradename
4507/45D
SciLabware
membrane size
0.2 μm
fitting
GL 45
General description
  • Polypropylene screwcap with integral 0.2 um PTFE membrane
  • Gas permeable membrane permits pressure equalisation allowing glassware to be sterilised using an autoclave with the screwcap fastened
  • Provides a tight seal that prevents the ingress of liquids or solids
  • Ideal for aerobic microbiology and cell culture
  • Suitable for storage and transport of gas generating media
  • Autoclavable
  • Temperature resistant to 140 deg. C
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)