Skip to Content
Merck
All Photos(1)

Key Documents

MT51350100

Mettler-Toledo protein cleaning solution

volume 250 mL

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€28.80

€28.80


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
€28.80

About This Item

UNSPSC Code:
41115600
NACRES:
NB.79

€28.80


Please contact Customer Service for Availability

form

liquid

manufacturer/tradename

Mettler-Toledo 51350100

volume

250 mL

General description

Pepsin-HCl for cleaning junctions with protein contamination. Treatment time about 1 h.

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service