MT51343102
Mettler-Toledo InLab® Expert pH electrode
with integrated temperature sensor, BNC/RCA (Cinch) connector
Select a Size
€402.00
Select a Size
About This Item
€402.00
Recommended Products
material
PEEK shaft
glass, U membrane
description
Reference electrolyte XEROLYT™ Polymer
Reference system ARGENTHAL™
feature
BNC/RCA (Cinch) connector
manufacturer/tradename
Mettler-Toledo 51343102
temperature measuring range
0-100 °C
L × diam.
120 mm × 12 mm
cable L
2 m
pH range
0-14
Looking for similar products? Visit Product Comparison Guide
General description
Ideal for a wide range of applications including emulsions, suspensions, effluent water and solutions containing proteins and sulphides.
The InLab Expert electrode has been designed with long life in mind.
The fine-pored ceramic junction used in conventional electrodes can easily become contaminated in critical media such as suspensions and wastewater. In contrast the InLab Expert electrode with a solid polymeric electrolyte requires no junction at all. As a result these sensors are particularly insensitive to contamination.
The reference electrode on these sensors is in direct contact with the measured medium by means of two open junctions, hence there is no junction to block. In addition, the high grade plastic (PEEK) shaft offers a high level of chemical resistance and robustness. Service life and measurement accuracy are further improved by the ARGENTHAL™ reference system which reduces loss of silver chloride from the reference wire at differing sample temperatures. Different cables can be connected to the MultiPin™ head allowing use with a wide range of meters.
- High grade plastic shaft (PEEK)
- Polymer electrolyte XEROLYT™
- Open junction
- ARGENTHAL reference
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service