MT30671568
Mettler Toledo SevenDirect SD23 pH / Conductivity Meter Pure H2O Kit
electrode, with InLab® Pure Pro-ISM & InLab® 741-ISM
About This Item
Recommended Products
AC/DC input
, universal plug set
220 V / 240 V
feature
electrode with InLab® Pure Pro-ISM & InLab® 741-ISM
packaging
pack of 1 kit
manufacturer/tradename
Mettler-Toledo 30671568
conductivity measuring range
0.001-2000 mS/cm, accuracy: ±0.5%, resolution: 0.001-2
mV measuring range
-2,000.000-2,000.000 mV, resolution: 1; 0.1
pH measuring range
-2.000-20.000, accuracy: ±0.002, resolution: 0.1
resolution: 0.001
resolution: 0.01
temperature measuring range
-30-130 °C, resolution: 0.1 °C
General description
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
Pure water specialist sensors
InLab Pure-Pro ISM and InLab 741-ISM are specialist sensors for pH, temperature and conductivity measurements in pure water. They are specifically designed to achieve high accuracy in this challenging sample.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
Two Parameters in One Device
Measure pH and conductivity with one instrument - simultaneously or separately as the application requires.
Application
Features and Benefits
- Large color touchscreen provides direct measurement and easy data entry.
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- Smart design and cover keep the instrument safe from dust and spills
- Combination of specialist pH, temperature and conductivity sensors make them highly suited for measurements in pure water.
- Electrode arm for efficient operation
- Protect critical settings Protected with two-level user management
- Display adapts to your needs to maximize readability
- Measure pH and conductivity with one instrument - simultaneously or separately.
Components
Legal Information
Kit Components Only
- Includes: Meter;Sensor; an efficient electrode arm; power supply; pH buffer sachets for initial calibration.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service