Skip to Content
Merck
All Photos(1)

Key Documents

D12600

Sigma-Aldrich

3,4-Diaminobenzoic acid

97%

Synonym(s):

4-Carboxy-o-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H3CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
775892
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

208-210 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Nc1ccc(cc1N)C(O)=O

InChI

1S/C7H8N2O2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H,8-9H2,(H,10,11)

InChI key

HEMGYNNCNNODNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Diaminobenzoic acid can be used to prepare:
  • Schiff base derivatives by reacting with substituted aldehydes and their corresponding metal complexes.
  • Poly(2,5-benzimidazole) (ABPBI) polymer by reacting with methanesulfonic acid and P2O5.
  • Pt-based Schiff base complexes applicable in H2O splitting reactions.

3,4-Diaminobenzoic acid undergoes cyclocondensations to form, for example, quinoxalines and benzimidazoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Sundaraganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 70(2), 376-383 (2007-12-28)
The Fourier Transform Raman and Fourier Transform infrared spectra of 3,4-diaminobenzoic acid (3,4-DABA) were recorded in the solid phase. Geometry optimizations were done without any constraint and harmonic-vibrational wave numbers and several thermodynamic parameters were calculated for the minimum energy
New platinum and ruthenium Schiff base complexes for water splitting reactions
Wang C, et al.
Dalton Transactions, 44(32), 14483-14493 (2015)
L A Cooper et al.
Antonie van Leeuwenhoek, 50(1), 53-62 (1984-01-01)
The effect of various compounds on growth, melanin biosynthesis and cell differentiation was studied in a hyaline (SH25) and a pigmented (SH25B) strain of Microdochium bolleyi. Dark pigment production by the hyaline strain was induced by the presence of DOPA
S Ram et al.
Journal of medicinal chemistry, 35(3), 539-547 (1992-02-07)
A series of methyl and ethyl 5-(alkoxycarbonyl)-1H-benzimidazole-2-carbamates (7-19) and methyl 5-carbamoyl-1H-benzimidazole-2-carbamates (24-34) have been synthesized via the reaction of an appropriate alcohol or amine with the acid chloride derivatives 6a or 6b at room temperature. Reaction of an alcohol with
Tetrahedron, 49, 9823-9823 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service