CLS9944916
Pyrex® disposable culture tubes, screw cap style, round-bottom
capacity 11.5 mL
Synonym(s):
culture tube, glass tubes, test tubes, Corning® 99449
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
borosilicate glass
round bottom tube
thread size
15-415
sterility
non-sterile
feature
cap: no
packaging
pkg of 1000 ea
pkg of 250 ea
manufacturer/tradename
Corning 99449-16
O.D. × L
16 mm × 100 mm
capacity
11.5 mL
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
- Pyrex® disposable culture tubes are made of low-expansion borosilicate glass.
- Ideal for use in tissue culture and general bacteriological applications.
- Screw-cap tubes are supplied without caps.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service