CLS9944516
Pyrex® disposable culture tubes, rimless
capacity 15.0 mL
Synonym(s):
culture tube, glass tubes, test tubes, Corning® 99445
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
round bottom tube
description
bulk packed
sterility
non-sterile
feature
cap: no
packaging
pkg of 1000 ea
manufacturer/tradename
Corning 99445-16
O.D. × L
16 mm × 100 mm
capacity
15.0 mL
color
clear
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Single-use culture tubes are convenience packed.
Application
Pyrex® disposable culture tubes, rimless have been used in:
- solid phase extraction (SPE)
- in the enrichment of phages that infect Marinomonas mediterranea
Features and Benefits
- Packed in convenient bulk packs that are simple to open and use
- Made up of borosilicate glass that minimizes pH changes and potential contaminants excreted from soda-lime glass
- Product does not contain animal-derived materials
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Estrogen-like effects in male goldfish co-exposed to fluoxetine and 17 alpha-ethinylestradiol
Environmental Science & Technology, 47, 5372-5382 (2013)
Isolation of phages infecting Marinomonas mediterranea by an enrichment protocol
Bio-protocol, e2921-e2921 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service