BR87818
BRAND® PP beaker with spout, low form
volume 600 mL
Synonym(s):
Beakers, Plastics
All Photos(2)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Recommended Products
material
polypropylene
description
in addition to ISO 7056
feature
graduations
beaded rim and spout
manufacturer/tradename
BRAND 87818
volume
600 mL
General description
PP, high clarity. ISO 7056. With graduation, beaded rim and spout. Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions.
Other Notes
Browse all plastic beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)